Assay Details
| Gene Symbol: eif-2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062713.6 | NP_495114.3 | 4 - 5 | 1542 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif-2D| Gene Symbol: eif-2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062713.6 | NP_495114.3 | 4 - 5 | 1542 | 134 | |
Set Membership: |
|