Assay Details
| Gene Symbol: C18A3.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062724.7 | NP_495125.1 | 3 - 4 | 401 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C18A3.3| Gene Symbol: C18A3.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062724.7 | NP_495125.1 | 3 - 4 | 401 | 124 | |
Set Membership: |
|