Assay Details
| Gene Symbol: cpb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062835.5 | NP_495236.3 | 5 - 6 | 1027 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpb-2| Gene Symbol: cpb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062835.5 | NP_495236.3 | 5 - 6 | 1027 | 65 | |
Set Membership: |
|