Assay Details
| Gene Symbol: tat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383859.1 | 13 - 14 | 3066 | 119 | ||
| NR_165322.1 | 13 - 14 | 3012 | 119 | |||
| NR_165323.1 | 13 - 14 | 3012 | 119 | |||
More Information
Set Membership: |
|

tat-4
Genomic Map