Assay Details
| Gene Symbol: tat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383859.1 | 4 - 5 | 998 | 95 | ||
| NR_165322.1 | 4 - 5 | 944 | 95 | |||
| NR_165323.1 | 4 - 5 | 944 | 95 | |||
More Information
Set Membership: |
|

tat-4
Genomic Map