Assay Details
| Gene Symbol: dgk-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368593.5 | NP_001355400.1 | 4 | 322 | 79 | |
| NM_001368594.2 | NP_001355399.1 | 2 - 3 | 288 | 79 | ||
| NM_062900.4 | NP_495301.1 | 2 - 3 | 180 | 79 | ||
More Information
Set Membership: |
|

dgk-5
Genomic Map