Assay Details
| Gene Symbol: dgk-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368593.5 | NP_001355400.1 | 10 | 1773 | 79 | |
| NM_001368594.2 | NP_001355399.1 | 8 - 9 | 1739 | 79 | ||
| NM_001368595.4 | NP_001355416.1 | 6 | 1440 | 79 | ||
| NM_001368596.3 | NP_001355417.1 | 6 - 7 | 2265 | 79 | ||
| NM_062900.4 | NP_495301.1 | 8 - 9 | 1631 | 79 | ||
More Information
Set Membership: |
|

dgk-5
Genomic Map