Assay Details
| Gene Symbol: dgk-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368593.5 | NP_001355400.1 | 11 | 1927 | 80 | |
| NM_001368594.2 | NP_001355399.1 | 9 - 10 | 1893 | 80 | ||
| NM_001368595.4 | NP_001355416.1 | 7 | 1594 | 80 | ||
| NM_001368596.3 | NP_001355417.1 | 7 - 8 | 2419 | 80 | ||
| NM_062900.4 | NP_495301.1 | 9 - 10 | 1785 | 80 | ||
More Information
Set Membership: |
|

dgk-5
Genomic Map