Assay Details
| Gene Symbol: sra-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062927.3 | NP_495328.2 | 1 - 2 | 267 | 177 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sra-27| Gene Symbol: sra-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062927.3 | NP_495328.2 | 1 - 2 | 267 | 177 | |
Set Membership: |
|