Assay Details
| Gene Symbol: sra-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062928.2 | NP_495329.1 | 2 - 3 | 650 | 182 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sra-28| Gene Symbol: sra-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062928.2 | NP_495329.1 | 2 - 3 | 650 | 182 | |
Set Membership: |
|