Assay Details
| Gene Symbol: klf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027033.2 | NP_001022204.1 | 2 - 3 | 532 | 71 | |
| NM_001027034.5 | NP_001022205.1 | 4 - 5 | 724 | 71 | ||
More Information
Set Membership: |
|

klf-3
Genomic Map