Assay Details
| Gene Symbol: klf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027033.2 | NP_001022204.1 | 3 - 4 | 702 | 67 | |
| NM_001027034.5 | NP_001022205.1 | 5 - 6 | 894 | 67 | ||
More Information
Set Membership: |
|

klf-3
Genomic Map