Assay Details
| Gene Symbol: ift-74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356863.3 | NP_001343748.1 | 1 - 2 | 131 | 83 | |
| NM_062958.6 | NP_495359.2 | 2 - 3 | 376 | 83 | ||
More Information
Set Membership: |
|

ift-74
Genomic Map