Assay Details
| Gene Symbol: ift-74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356863.3 | NP_001343748.1 | 3 - 4 | 528 | 99 | |
| NM_062958.6 | NP_495359.2 | 4 - 5 | 773 | 99 | ||
More Information
Set Membership: |
|

ift-74
Genomic Map