Assay Details
| Gene Symbol: agr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026981.6 | NP_001022152.3 | 20 - 21 | 3220 | 126 | |
| GenBank mRNA | AM773423.1 | - | 20 - 21 | 3220 | 126 | |
More Information
Set Membership: |
|

agr-1
Genomic Map