Assay Details
| Gene Symbol: pmp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063007.5 | NP_495408.1 | 6 - 7 | 1035 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pmp-2| Gene Symbol: pmp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063007.5 | NP_495408.1 | 6 - 7 | 1035 | 108 | |
Set Membership: |
|