Assay Details
| Gene Symbol: hrpl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063010.6 | NP_495411.2 | 4 - 5 | 417 | 79 | |
| NM_063011.6 | NP_495412.1 | 3 - 4 | 289 | 79 | ||
| Gene Symbol: psmd-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383874.1 | 3 - 4 | 305 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383874.1 transcripts.
Set Membership: |
|

hrpl-1
Genomic Map