Assay Details
| Gene Symbol: hrpl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063010.6 | NP_495411.2 | 9 - 10 | 1139 | 122 | |
| Gene Symbol: psmd-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383874.1 | 8 - 9 | 1027 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383874.1 transcripts.
Set Membership: |
|

hrpl-1
Genomic Map