Assay Details
| Gene Symbol: cyn-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026719.5 | NP_001021890.1 | 2 - 3 | 243 | 102 | |
| NM_063015.5 | NP_495416.2 | 2 - 3 | 245 | 102 | ||
| GenBank mRNA | U34954.1 | - | 2 - 3 | 242 | 102 | |
More Information
Set Membership: |
|

cyn-10
Genomic Map