Assay Details
| Gene Symbol: flcn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026937.4 | NP_001022108.1 | 1 - 2 | 249 | 68 | |
| NM_001381470.1 | NP_001368185.1 | 1 - 2 | 88 | 68 | ||
More Information
Set Membership: |
|

flcn-1
Genomic Map