Assay Details
| Gene Symbol: flcn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026937.4 | NP_001022108.1 | 2 - 3 | 354 | 108 | |
| NM_001276732.2 | NP_001263661.1 | 14 - 15 | 1367 | 108 | ||
| NM_001306549.3 | NP_001293478.1 | 14 - 15 | 1358 | 108 | ||
| NM_001306551.2 | NP_001293480.1 | 14 - 15 | 1379 | 108 | ||
| NM_001330952.3 | NP_001317868.1 | 14 - 15 | 1377 | 108 | ||
| NM_001381470.1 | NP_001368185.1 | 2 - 3 | 193 | 108 | ||
| GenBank mRNA | HE963850.1 | - | 13 - 14 | 1273 | 108 | |
More Information
Set Membership: |
|

flcn-1
Genomic Map