Assay Details
| Gene Symbol: flcn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026937.4 | NP_001022108.1 | 3 - 4 | 480 | 67 | |
| NM_001276732.2 | NP_001263661.1 | 15 - 16 | 1493 | 67 | ||
| NM_001306549.3 | NP_001293478.1 | 15 - 16 | 1484 | 67 | ||
| NM_001306551.2 | NP_001293480.1 | 15 - 16 | 1505 | 67 | ||
| NM_001330952.3 | NP_001317868.1 | 15 - 16 | 1503 | 67 | ||
| NM_001381470.1 | NP_001368185.1 | 3 - 4 | 319 | 67 | ||
| GenBank mRNA | HE963850.1 | - | 14 - 15 | 1399 | 67 | |
More Information
Set Membership: |
|

flcn-1
Genomic Map