Assay Details
| Gene Symbol: flcn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276732.2 | NP_001263661.1 | 11 - 12 | 1110 | 78 | |
| NM_001306549.3 | NP_001293478.1 | 11 - 12 | 1101 | 78 | ||
| NM_001306551.2 | NP_001293480.1 | 11 - 12 | 1110 | 78 | ||
| NM_001330952.3 | NP_001317868.1 | 11 - 12 | 1108 | 78 | ||
| GenBank mRNA | HE963850.1 | - | 10 - 11 | 1004 | 78 | |
More Information
Set Membership: |
|

flcn-1
Genomic Map