Assay Details
| Gene Symbol: eif-2Bgamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063027.8 | NP_495428.1 | 6 - 7 | 938 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif-2Bgamma| Gene Symbol: eif-2Bgamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063027.8 | NP_495428.1 | 6 - 7 | 938 | 96 | |
Set Membership: |
|