Assay Details
| Gene Symbol: lsm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063113.6 | NP_495514.1 | 3 - 4 | 285 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lsm-4| Gene Symbol: lsm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063113.6 | NP_495514.1 | 3 - 4 | 285 | 129 | |
Set Membership: |
|