Assay Details
| Gene Symbol: brf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322739.3 | NP_001309557.1 | 6 - 7 | 2049 | 130 | |
| NM_001322740.4 | NP_001309558.1 | 6 - 7 | 2037 | 130 | ||
More Information
Set Membership: |
|

brf-1
Genomic Map