Assay Details
| Gene Symbol: insc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381495.1 | NP_001368642.1 | 7 - 8 | 748 | 89 | |
| NM_001381496.2 | NP_001367460.1 | 7 - 8 | 772 | 89 | ||
More Information
Set Membership: |
|

insc-1
Genomic Map