Assay Details
| Gene Symbol: ubc-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047290.4 | NP_001040755.1 | 3 - 4 | 670 | 71 | |
| NM_001047291.5 | NP_001040756.1 | 2 - 3 | 275 | 71 | ||
More Information
Set Membership: |
|

ubc-6
Genomic Map