Assay Details
| Gene Symbol: tph-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373740.3 | NP_001359899.1 | 1 | 73 | 89 | |
| NM_063183.6 | NP_495584.4 | 1 - 2 | 115 | 89 | ||
| GenBank mRNA | AF135186.1 | - | 1 - 2 | 135 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001373740.3 transcripts.
Set Membership: |
|

tph-1
Genomic Map