Assay Details
| Gene Symbol: tph-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063183.6 | NP_495584.4 | 2 - 3 | 211 | 89 | |
| GenBank mRNA | AF135186.1 | - | 2 - 3 | 231 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tph-1| Gene Symbol: tph-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063183.6 | NP_495584.4 | 2 - 3 | 211 | 89 | |
| GenBank mRNA | AF135186.1 | - | 2 - 3 | 231 | 89 | |
Set Membership: |
|