Assay Details
| Gene Symbol: tph-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373740.3 | NP_001359899.1 | 3 | 669 | 62 | |
| NM_063183.6 | NP_495584.4 | 4 - 5 | 671 | 62 | ||
| GenBank mRNA | AF135186.1 | - | 4 - 5 | 691 | 62 | |
More Information
Set Membership: |
|

tph-1
Genomic Map