Assay Details
| Gene Symbol: tph-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373740.3 | NP_001359899.1 | 6 | 930 | 137 | |
| NM_063183.6 | NP_495584.4 | 7 - 8 | 932 | 137 | ||
| GenBank mRNA | AF135186.1 | - | 7 - 8 | 952 | 137 | |
More Information
Set Membership: |
|

tph-1
Genomic Map