Assay Details
| Gene Symbol: tph-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373740.3 | NP_001359899.1 | 8 | 1189 | 75 | |
| NM_063183.6 | NP_495584.4 | 9 - 10 | 1191 | 75 | ||
| GenBank mRNA | AF135186.1 | - | 9 - 10 | 1211 | 75 | |
More Information
Set Membership: |
|

tph-1
Genomic Map