Assay Details
| Gene Symbol: cpna-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063221.5 | NP_495622.2 | 11 - 12 | 2092 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpna-5| Gene Symbol: cpna-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063221.5 | NP_495622.2 | 11 - 12 | 2092 | 68 | |
Set Membership: |
|