Assay Details
| Gene Symbol: B0228.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063226.5 | NP_495627.3 | 3 - 4 | 462 | 147 | |
| Gene Symbol: trx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026715.6 | NP_001021886.1 | 2 - 3 | 272 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001026715.6 transcripts.
Set Membership: |
|

B0228.6
Genomic Map