Assay Details
| Gene Symbol: cki-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063240.5 | NP_495641.1 | 1 - 2 | 225 | 89 | |
| GenBank mRNA | AF179358.1 | - | 1 - 2 | 247 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cki-1| Gene Symbol: cki-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063240.5 | NP_495641.1 | 1 - 2 | 225 | 89 | |
| GenBank mRNA | AF179358.1 | - | 1 - 2 | 247 | 89 | |
Set Membership: |
|