Assay Details
| Gene Symbol: ptc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373748.2 | NP_001360447.1 | 4 - 5 | 521 | 117 | |
| NM_063261.5 | NP_495662.2 | 3 - 4 | 339 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001373748.2 transcripts.
Set Membership: |
|

ptc-1
Genomic Map