Assay Details
| Gene Symbol: sfxn-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026851.6 | NP_001022022.1 | 1 - 2 | 144 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sfxn-5| Gene Symbol: sfxn-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026851.6 | NP_001022022.1 | 1 - 2 | 144 | 139 | |
Set Membership: |
|