Assay Details
| Gene Symbol: ire-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267206.3 | NP_001254135.1 | 6 - 7 | 957 | 121 | |
| GenBank mRNA | AF435952.1 | - | 5 - 6 | 820 | 121 | |
More Information
Set Membership: |
|

ire-1
Genomic Map