Assay Details
| Gene Symbol: ire-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267206.3 | NP_001254135.1 | 9 - 10 | 2803 | 90 | |
| NM_001267207.3 | NP_001254136.1 | 3 - 4 | 1628 | 90 | ||
| GenBank mRNA | AF435952.1 | - | 8 - 9 | 2666 | 90 | |
More Information
Set Membership: |
|

ire-1
Genomic Map