Assay Details
| Gene Symbol: emb-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063311.9 | NP_495712.1 | 6 - 7 | 1764 | 111 | |
| GenBank mRNA | AF314467.1 | - | 6 - 7 | 1763 | 111 | |
More Information
Set Membership: |
|

emb-27
Genomic Map