Assay Details
| Gene Symbol: qua-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063324.4 | NP_495725.2 | 5 - 6 | 610 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

qua-1| Gene Symbol: qua-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063324.4 | NP_495725.2 | 5 - 6 | 610 | 91 | |
Set Membership: |
|