Assay Details
| Gene Symbol: dab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129154.6 | NP_001122626.2 | 2 - 3 | 211 | 61 | |
| NM_001373756.2 | NP_001360462.1 | 2 - 3 | 211 | 61 | ||
| NM_001383903.1 | 1 - 2 | 121 | 61 | |||
| NM_063329.8 | NP_495730.3 | 2 - 3 | 211 | 61 | ||
More Information
Set Membership: |
|

dab-1
Genomic Map