Assay Details
| Gene Symbol: M110.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063333.4 | NP_495734.3 | 9 - 10 | 1297 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

M110.7| Gene Symbol: M110.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063333.4 | NP_495734.3 | 9 - 10 | 1297 | 151 | |
Set Membership: |
|