Assay Details
| Gene Symbol: nlp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063334.6 | NP_495735.1 | 4 - 5 | 327 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nlp-5| Gene Symbol: nlp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063334.6 | NP_495735.1 | 4 - 5 | 327 | 103 | |
Set Membership: |
|