Assay Details
| Gene Symbol: nst-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063348.6 | NP_495749.1 | 4 - 5 | 1085 | 72 | |
| Gene Symbol: mrpl-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063346.6 | NP_495747.1 | 4 - 5 | 1074 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_063346.6 transcripts.
Set Membership: |
|

nst-1
Genomic Map