Assay Details
| Gene Symbol: drd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129140.4 | NP_001122612.1 | 4 - 5 | 460 | 179 | |
| NM_063362.6 | NP_495763.1 | 4 - 5 | 745 | 179 | ||
| NR_023951.1 | 4 - 5 | 452 | 179 | |||
More Information
Set Membership: |
|

drd-1
Genomic Map