Assay Details
| Gene Symbol: skpo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063367.7 | NP_495768.1 | 19 - 20 | 4330 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

skpo-1| Gene Symbol: skpo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063367.7 | NP_495768.1 | 19 - 20 | 4330 | 120 | |
Set Membership: |
|