Assay Details
| Gene Symbol: sra-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027015.1 | NP_001022186.1 | 1 - 2 | 247 | 111 | |
| NM_001027016.3 | NP_001022187.1 | 1 - 2 | 292 | 111 | ||
More Information
Set Membership: |
|

sra-13
Genomic Map