Assay Details
| Gene Symbol: acp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063375.2 | NP_495776.1 | 6 - 7 | 750 | 175 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acp-3| Gene Symbol: acp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063375.2 | NP_495776.1 | 6 - 7 | 750 | 175 | |
Set Membership: |
|